Ammonium sulfate 99.0-100.5%, granules ACS, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = RL
BAKR0795-08
BAKR0795-12
BAKR0795-65
CAJT0795-8EA
7866.09
CAD
CAJT0795-8
CAJT0795-12
CAJT0795-65
Ammonium sulfate 99.0-100.5%, granules ACS, FCC, J.T.Baker®
Ammonium sulphate
Food GMP manufactured product.
Formula:
(NH₄)₂SO₄ Melting Pt: 336…339 °C Density: 1.78 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
ACS - Assay ((NH₄)₂SO₄) | 99.0 - 100.5 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Residue on Ignition (ACS) | ≤0.005 % |
ACS - pH of 5% Solution at 25°C | 5.0 - 6.0 |
ACS - Nitrate (NO₃) | ≤0.001 % |
Chloride (Cl) | ≤3 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Phosphate (PO₄) | ≤3 ppm |
Heavy Metals (as Pb) | ≤3 ppm |
FCC - Assay ((NH₄)₂SO₄)(F.C.C.) | 99.0 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Residue on Ignition (FCC) | ≤0.25 % |
FCC - Lead (Pb) | ≤3 mg/kg |
FCC - Selenium (Se) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...