β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate, 2-Morpholinoethanesulfonic acid hydrate, 2-(N-Morpholino)ethanesulfonic acid monohydrate

Management of Change (MOC) category = NI

BAKR4812-02 BAKR4812-04 BAKR4812-06 BAKR4812-08
CAJT4812-2EA 3436.28 CAD
CAJT4812-2 CAJT4812-4 CAJT4812-6 CAJT4812-08
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate

Suitable for use in biopharmaceutical manufacturing applications.

Formula: C₆H₁₃NO₄S·1H₂O
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Identification (by IR) Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Color of a 1M Alkaline Solution (Colorless) Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR