D-Lactose monohydrate, powder NF, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
D-(+)-Lactose monohydrate
Management of Change (MOC) category = R
BAKR2250-07
BAKR2250-05
CAJT2250-7EA
2125.83
CAD
CAJT2250-7
CAJT2250-5
D-Lactose monohydrate, powder NF, Multi-Compendial
D-Lactose monohydrate
White to off-white powder.
Formula:
C₁₂H₂₂O₁₁·1H₂O MW: 360.32 g/mol Melting Pt: 223 Storage Temperature: Ambient |
MDL Number:
MFCD00150747 CAS Number: 64044-51-5 EINECS: 200-559-2 |
Specification Test Results
Meets J.P. Chemical Specifications | |
Meets N.F. Requirements | |
GMP Manufactured Product | |
Meets E.P. Chemical Specifications | |
Store in airtight container. | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Clarity and Color of Solution | Passes Test |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Specific Rotation [Å]²º^D (+) | 54.4 - 55.9 Degree |
NF - Microbial Limits - Total Aerobic Microbial Counts, cfu/g | ≤100 |
NF - Microbial Limits - Total Combined Molds and Yeasts Count (per g) | ≤50 |
NF - Microbial Limits - Escherichia Coli and Salmonella | Passes Test |
NF - Acidity or Alkalinity | Passes Test |
NF - Loss on Drying at 80°C | ≤0.5 % |
NF - Water (H₂O) | 4.5 - 5.5 % |
NF - Residue on Ignition | ≤0.1 % |
NF - Protein and Light-Absorbing Impurities | Passes Test |
EP - Identification A | Passes Test |
EP - Identification B | Passes Test |
EP - Identification C | Passes Test |
EP - Identification D | Passes Test |
EP - Specific Rotation [Å]²º^D (+) | 54.4 - 55.9 Degree |
EP - Sulfated Ash | ≤0.1 % |
EP - Microbial Limits - Total Aerobic Microbial Count, cfu/g | ≤100 |
EP- Microbial Limits - Total Combined Molds and Yeasts Count (per g) | ≤50 |
EP - Microbial Limits - Escherichia coli (absent) | Passes Test |
EP - Acidity or Alkalinity | Passes Test |
EP - Water (H₂O) | 4.5 - 5.5 % |
EP - Protein and Light-Absorbing Impurities | Passes Test |
EP - Appearance of Solution | Passes Test |
JP - Clarity and Color of Solution | Passes Test |
JP - Identification A | Passes Test |
JP - Specific Rotation [Å]²º^D (+) | 54.4 - 55.9 Degree |
JP - Microbial Limits - Total Aerobic Microbial Count, cfu/g | ≤100 |
JP - Microbial Limits - Total Combined Molds and Yeasts Count (per g) | ≤50 |
JP - Microbial Limits - Escherichia Coli and Salmonella | Passes Test |
JP - Acidity or Alkalinity | Passes Test |
JP - Water (H₂O) | 4.5 - 5.5 % |
JP - Loss on Drying at 80°C | ≤0.5 % |
JP - Residue on Ignition | ≤0.10 % |
JP - Heavy Metals (as Pb) | ≤5 ppm |
JP - Protein and Light-Absorbing Impurities | Passes Test |
Endotoxin Concentration (EU/g), For Information Only | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. | |
Internationally Harmonized |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...