L(+)-Valine 98.5-101.5% (dried basis) USP, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
H-Val-OH, Valine, (S)-(+)-Valine
Management of Change (MOC) category = R
BAKR2095-05
BAKR2095-06
CAJT2095-5EA
322.08
CAD
CAJT2095-5
CAJT2095-6
CAJT2095-7
L(+)-Valine 98.5-101.5% (dried basis) USP, Multi-Compendial
L(+)-Valine
White crystals.
- Food GMP manufactured product
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 217 Melting Pt: 86 Density: 1.284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064220 CAS Number: 72-18-4 EINECS: 200-773-6 Merck Index: 13,09458 |
Specification Test Results
Meets E.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
Meets J.P. Chemical Specifications | |
Meets B.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₅H₁₁NO₂) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D (+) | 26.6 - 28.8 Degree |
USP - pH | 5.5 - 7.0 |
USP - Loss on Drying at 105°C | ≤0.3 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Chloride (Cl) | ≤0.05 % |
USP - Sulfate (SO₄) | ≤0.03 % |
USP - Iron (Fe) | ≤0.003 % |
USP - Related Compounds - Individual Impurities | ≤0.5 % |
USP - Related Compounds - Total Impurities | ≤2.0 % |
EP/BP - Assay (C₅H₁₁NO₂) (dried basis) | 98.5 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Specific Rotation [Å]²º^D (+) | 26.5 - 29.0 Degree |
EP/BP-Ninhydrin-Positive Substances-Impurity B | ≤0.4 % |
EP/BP-Ninhydrin-Positive Substances-Each | ≤0.2 % |
EP/BP-Ninhydrin-Positive Substances-Total Impurities | ≤1.0 % |
EP/BP - Chloride (Cl) | ≤200 ppm |
EP/BP - Sulfate (SO₄) | ≤300 ppm |
EP/BP - Ammonium (NH₄) | ≤0.02 % |
EP/BP - Iron (Fe) | ≤10 ppm |
EP/BP - Loss on Drying | ≤0.5 % |
EP/BP - Ash (sulfated) | ≤0.1 % |
JP - Assay (C₅H₁₁NO₂) (dried basis) | 98.5 - 101.0 % |
JP - Identification | Passes Test |
JP - Optical Rotation (+) | 26.5 - 29.0 Degree |
JP - pH | 5.5 - 6.5 |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤0.021 % |
JP - Sulfate (SO₄) | ≤0.028 % |
JP - Ammonium (NH₄) | ≤0.02 % |
JP - Heavy Metals (as Pb) | ≤20 ppm |
JP - Arsenic (As) | ≤2 ppm |
JP - Related Substances | Passes Test |
JP - Loss on Drying at 105°C | ≤0.30 % |
JP - Residue on Ignition | ≤0.10 % |
Endotoxin Concentration, IU/mg, For Information Only | |
Only Class 2 Solvents (Methanol) are likely to be present. All are | |
below Option 1 limits. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...