L(-)-Tryptophan 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = R
BAKR2092-06
BAKR2092-07
CAJT2092-6EA
4770.68
CAD
CAJT2092-6
CAJT2092-7
L(-)-Tryptophan 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
L(-)-Tryptophan
White crystals.
- Food GMP manufactured product
Formula:
C₁₁H₁₂N₂O₂ Melting Pt: 290 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Specification Test Results
GMP Manufactured Product | |
Meets J.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets U.S.P Requirements | |
Preserve in well-closed containers. | |
Store in well-closed,light-resistant containers | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₁₁H₁₂N₂O₂) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D | -32.8 - -29.4 Degree |
USP - pH | 5.5 - 7.0 |
USP - Loss on Drying at 105°C | ≤0.3 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Chloride (Cl) | ≤0.05 % |
USP - Sulfate (SO₄) | ≤0.03 % |
USP - Iron (Fe) | ≤0.003 % |
USP - Organic Impurities - Total Impurities 1 | ≤0.01 % |
USP - Organic Impurities - Total Impurities 2 | ≤0.03 % |
USP - Organic Impurities - Tryptophan Related Compound A | ≤10 ppm |
JP - Assay (C₁₁H₁₂N₂O₂) (dried basis) | 98.5 - 101.0 % |
JP - Identification | Passes Test |
JP - Specific Rotation [Å]²º^D | -33.0 - -30.0 Degree |
JP - pH | 5.4 - 6.4 |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤0.021 % |
JP - Sulfate (SO₄) | ≤0.028 % |
JP - Ammonium (NH₄) | ≤0.02 % |
JP - Heavy Metals (as Pb) | ≤20 ppm |
JP - Arsenic (As) | ≤2 ppm |
JP - Related Substances | Passes Test |
JP - Loss on Drying at 105°C | ≤0.30 % |
JP - Residue on Ignition | ≤0.10 % |
Endotoxin Concentration, IU/mg, For Information Only | |
EP - Identification A | Passes Test |
EP - Identification B | Passes Test |
EP - Appearance of Solution | Passes Test |
EP - Specific Optical Rotation [Å]²º^D | -33.0 - -30.0 Degree |
EP/BP-Ninhydrin-Positive Substances-Each | ≤0.2 % |
EP/BP-Ninhydrin-Positive Substances-Total Impurities | ≤0.5 % |
EP - 1,1'Ethylidenebistryptophan | ≤10 ppm |
EP - Sum of the Areas less than Tryptophan® | ≤100 ppm |
EP - Sum of the Areas greater than Tryptophan® | ≤300 ppm |
EP - Chloride (Cl) | ≤200 ppm |
EP - Sulfate (SO₄) | ≤300 ppm |
EP/BP - Ammonium (NH₄) | ≤0.02 % |
EP - Iron (Fe) | ≤20 ppm |
EP - Loss on Drying at 105°C | ≤0.5 % |
EP - Sulfated Ash | ≤0.1 % |
EP - Assay (C₁₁H₁₂N₂O₂) (dried basis) | 98.5 - 101.0 % |
Only Class 2 Solvents (Methanol) are likely to be present. All are | |
below Option 1 limits. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...