L(+)-Ascorbic acid 99, powder_fine USP 200-325 Mesh, multi-compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
M.O.C. Category R
BAKR0939-07
BAKR0939-08
CAJT0939-7EA
1538.76
CAD
CAJT0939-7
CAJT0939-8
L(+)-Ascorbic acid 99, powder_fine USP 200-325 Mesh, multi-compendial
L(+)-Ascorbic acid
White crystals.
- GMP manufactured product
Formula:
C₆H₈O₆ MW: 176.13 g/mol Boiling Pt: 553 Melting Pt: 190…192 °C Density: 1.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 EINECS: 200-066-2 REACH: exempt Merck Index: 13,00837 |
|
Specification Test Results
Meets J.P. Chemical Specifications | |
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₆H₈O₆) | 99.0 - 100.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D (+) | 20.5 - 21.5 Degree |
USP - Residue on Ignition | ≤0.1 % |
Mesh - Thru U.S. No. 100 Sieve | ≥100 % |
Mesh - Thru U.S. No. 200 Sieve | ≥95 % |
JP - Assay (C₆H₈O₆) (dried basis) | 99.0 - 101.0 % |
JP - Identification A | Passes Test |
JP - Identification B | Passes Test |
JP - Optical Rotation [Å]²º^D (+) | 20.5 - 21.5 Degree |
JP - pH (1 in 20) | 2.2 - 2.5 |
JP - Clarity and Color of Solution | Passes Test |
JP - Heavy Metals (as Pb) | ≤20 ppm |
JP - Loss on Drying | ≤0.20 % |
JP - Residue on Ignition | ≤0.10 % |
Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3 | |
Solvents are likely to be present. Residual Class 2 Solvents are below | |
Option 1 and the residual concentration of all Class 3 Solvents is | |
<0.5%. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...