Flip-Top Containers, Capitol Vial®
Supplier: Thermo Fisher Scientific
02CL 08CL 03CL 03CLYELLOW 03CLFREGORED
33500-728CS 386.11 CAD
33500-728 33999-042 46610-132 46610-134 46610-136 46610-138 46610-142 46610-144 46610-146 46610-152 46610-164 46610-172 46610-542 46610-544 46610-546 46610-548 46610-550 46610-552 46610-554 46610-556 46610-558 46610-560 46610-564 46610-566 46610-568 46610-570 46610-572 46610-576 46610-578 46610-580 46610-582 46610-588 46610-594
Flip-Top Containers, Capitol Vial®
Sample Containers General Purpose Containers
Triple-seal containers are airtight, leakproof, nonabsorptive, and nonaerosoling.
Unique process physically closes vials in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µm. Sterile.
Made of FDA-approved, food-grade 100% polypropylene. Hinged top permits opening and closing these flip-top vials with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing.
Certifications: Vials conform to FDA and SAMHSA specifications.
Ordering information: Container 46610-588 features 10mL increments.