Trifluoroacetic acid 0.1% (v/v) in acetonitrile mobile phase for HPLC, Supelco®

Supplier: MilliporeSigma

Synonyms: TFA

TX1278-NP20
CA-TX1278-NP20EA 2928.21 CAD
CA-TX1278-NP20
Trifluoroacetic acid 0.1% (v/v) in acetonitrile mobile phase for HPLC, Supelco®
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 1648
ADR: 3,II
https://ca.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Specification Test Results

Assay (trifluoroacetic acid) 0.045-0.055% (v/v)
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Gradient at 210 nm 0.005AU max
Gradient at 280 nm 0.001AU max
Residue after evaporation 5ppm max
UV Abs. at 190 nm 1.00AU max
UV Abs. at 200 nm 0.50AU max
UV Abs. at 210 nm 0.40AU max
UV Abs. at 230 nm 0.30AU max
UV Abs. at 250 nm 0.05AU max
UV Abs. at 280 nm 0.010AU max
UV Abs. at 300 nm 0.010AU max
UV Abs. at 350 nm 0.010AU max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR