Barnstead™ Hose Nipple Cartridges, Thermo Scientific
Supplier: Thermo Fisher Scientific
D8822 D8904 D8903 D8905 D8911 D8950 D8951 D8921 D8901 D8902 D8922
26303-258EA 269.99 CAD
26303-258 26303-562 26303-584 26303-595 26303-600 26303-653 26303-655 26303-711 26303-507 26303-530 26303-686
Barnstead™ Hose Nipple Cartridges, Thermo Scientific
Filters Filter Cartridges
Cartridge tubes and end caps are constructed of 100% virgin polypropylene and are designed for non pressure applications.
  • Cartridges include 9.5 mm (³/₈") barbed hose connections on each end
All cartridges contain dyes which change color when resin is exhausted, with the exception of the Organic Removal Cartridge.
Note: Non pressure refers to the inability of the cartridges to accept back pressure. Output water must be diverted to an atmospherically-vented receptacle.
Shipping weight: 2.3 kg (5 lbs.)