Barnstead Prefilter Cartridges, Thermo Scientific
Supplier: Thermo Fisher Scientific
18017 18018 18011 D2729
26303-041EA 120.62 CAD
26303-041 26303-043 26303-052 26303-879
Barnstead Prefilter Cartridges, Thermo Scientific
Filters Filter Cartridges
For removal of heavy particulates from feed water to extend final filter life.
  • Double open end
  • 10" length
For use in 26303-020 and 26303-176 filter holders.
Order Now
SPECIFICATIONS
Environmentally Preferable