VITLAB® Sample Containers with Screw Caps, PFA, BrandTech
Supplier: Brandtech
130297 130397 130497 130597
83008-658PK 275.31 CAD
83008-658 83008-663 83008-666 83008-670
VITLAB® Sample Containers with Screw Caps, PFA, BrandTech
Sample Containers General Purpose Containers
Containers and screw caps are made of PFA (perfluoroalkoxy copolymer), a fluoropolymer with excellent chemical resistance.
  • Trace analysis grade
  • Easy to clean
  • Screw on caps for security
Surfaces are much less porous than PTFE and are highly cleanable to reduce sample contamination by 'memory effect' from prior samples. Ideal for use in all aspects of ICP-MS and other trace analysis work. Containers provide excellent stability for standards and are manufactured without metal catalysts. Highly translucent for verification of contents. Thermally stable from –200 to 260 °C (–328 to 500 °F). Autoclavable.