Sample Tubes, 50 ml, Simport Scientific
Supplier: Simport Scientific
C571-2 C571-1
CA15705-376CS 373.56 CAD
CA15705-376 CA73960-336
Sample Tubes, 50 ml, Simport Scientific
Tubes Test Tubes
These polypropylene, conical bottom tubes with self-standing bases have molded-in graduations from 10 to 50 ml in 5 ml increments.
  • Chemical-resistant and shatterproof
  • With a leakproof screw cap, which is particularly important for transporting hazardous material
Ideal for transport and storage of urine, sputum, and most liquids or particulate samples.
Temperature: –90 to 121 °C (–130 to 250 °F).