Sieve Receiver Pans, W.S. Tyler™
Supplier: W.S. Tyler
8492 8494 8526 8481 8482 8484 8491
57332-100EA 248.94 CAD
57332-100 57332-102 57332-218 57355-006 57355-028 57355-040 57355-061
Sieve Receiver Pans, W.S. Tyler™
Trays/Dishes/Pots Laboratory Trays/Dishes
Sieve Receiver Pans are designed for use with 20.3cm (8") or 30.5cm (12") diameter sieves.
Pans with extended rims may be placed within a stack of sieves when sieving two or more samples simultaneously.