AcroPrep™ 96-Well Filter Plates, 1 ml, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
5063-N 5056-N 5053 5053 5041-N
CA16003-808PK 400.97 CAD
CA16003-808 CA28148-664 CA28148-626 28148-626 CA76894-024 CA-16003-806
AcroPrep™ 96-Well Filter Plates, 1 ml, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
Constructed of chemically resistant, biologically inert polypropylene, plates are optimized for a variety of applications requiring larger per well volume processing.
  • Chemically resistant material
  • Extended flow directors
  • Individually sealed wells
Plates feature extended flow directors to eliminate splattering and crosstalk of filtrate, a serialized bar code label for use in automated tracking systems, robotics-friendly design, standard SBS format, and individually sealed wells to eliminate crosstalk.
Available in a variety of membranes and membrane configurations to meet various application needs. Choose from GHP (hydrophilic polypropylene), glass fiber (borosilicate glass without binder), glass fiber over Bio-Inert® modified low protein-binding nylon, hydrophobic PTFE, Supor® polyethersulfone, or Omega™ polyethersulfone (PES) membranes.