Pierce™ ELISA Reagent Reservoirs, PS, Thermo Scientific
Supplier: Thermo Scientific
15075
CAPI15075PK 481.62 CAD
CAPI15075
Pierce™ ELISA Reagent Reservoirs, PS, Thermo Scientific
Reagent Reservoirs
Disposable reagent reservoirs molded from high-impact PS.
  • Sterile and durable
  • Reservoirs facilitate repeated pick-up of liquid reagents by multi-channel pipettors for delivery to 96-well plates