Flip-Top Containers with Lock Seal, Capitol Vial®
Supplier: Thermo Fisher Scientific
04HPLS 02LS
46610-584CS 395.4 CAD
46610-584 46610-586 46610-596 46610-590
Flip-Top Containers with Lock Seal, Capitol Vial®
Sample Containers General Purpose Containers
Triple-seal containers are sterile, airtight, and leakproof
Unique process physically closes containers in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µ. Process also produces sterile containers that do not require a second means of sterilization for most applications. Color: clear.
Nonabsorptive, nonaerosoling containers are made of FDA-approved, food grade 100% polypropylene. Hinged top enables opening and closing with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging,More Product Information