DUOSEAL® Belt-Drive Vacuum Pumps, Single Stage, Welch®
Supplier: Welch Vacuum
1399B-01 1380B-01
54972-709EA 5956.63 CAD
54972-709 54987-203
DUOSEAL® Belt-Drive Vacuum Pumps, Single Stage, Welch®
Pumps Vacuum Pumps
Pumps are ideal for continuous-duty applications, including vacuum distillations, glove box transfer chamber, residue solvent removal, and resin degassing
They feature thermal overload protection standard and operate at a low rpm which allows them to reach a low ultimate pressure (0.02mm Hg). Operating at low speed also reduces the rate of friction and wear on pump components which ensures a long service life.
Ordering information: Exhaust filters are available separately and are supplied with housing and element.