Multi-Purpose Sampling Cups, Globe Scientific
Supplier: Globe Scientific
110811 110711 110021 110610 110621
60820-062PK 137.24 CAD
60820-062 60820-054 60820-056 60820-058 60820-060
Multi-Purpose Sampling Cups, Globe Scientific
Sample Containers General Purpose Containers
Transparent, polystyrene cups are durable and chemical-resistant
  • Polystyrene (PS)
  • Can be used on most common brands of instruments
They feature a wide base design to prevent tipping or spilling and a conical bottom for use in microsampling. A variety of cup sizes permits use with most autoanalyzers. Cups are designed to accept snap caps (not included).