BD BBL™ Microbiological Specimen Collection and Transport Systems, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
220118 220115 220099 220117 220134 220105 220133 220109 220130 220099 220132 220121 220135 220122 220133 220105 220117 220126 220122 220119 220123 220097 220116 220115 220123 220126 220093 220129 220134 220116 220131 220119 220125 220118 220093 220109 220132 220125 BBL-220121 220121 220097
CA90001-070PK 329.37 CAD
90001-080 90001-082 90001-084 90001-040 CA90001-070 90001-062 CA90001-050 90001-086 90001-064 CA90001-072 90001-044 90001-066 CA90001-052 CA90001-054 90001-046 CA90001-046 90001-058 90001-036 90001-038 CA-90001-082 CA-90001-084 90001-070 90001-050 90001-072 CA90001-040 CA90001-062 90001-074 90001-052 90001-054 90001-076 CA90001-064 CA90001-044 CA90001-066 90001-078 90001-056 CA-90001-080 76240-580 CA90001-056 CA90001-036 CA90001-058 CA90001-038
BD BBL™ Microbiological Specimen Collection and Transport Systems, BD Diagnostics
Swabs Transport Swabs
Keep samples viable during transport with convenient, self-contained BD CultureSwab™ and BD CultureSwab Plus™ systems
Each BD CultureSwab™ tube contains a swab with a polyurethane sponge. After sampling, the swab is returned to the tube and specimen is put in immediate contact with the medium and kept moist during transport.
Each BD CultureSwab Plus™ tube contains a 5mL column of Amies gel for maximum specimen protection and improved sample viability of anaerobes. Gel column also ensures that swab maintains a centralized position during transport. BD CultureSwab Plus™ tubes feature a special venturi hourglass design that helps prevent breakdown of the gel column during transport, minimizes bubbles and breaks in gel, and reduces the formation of air pockets that are harmful to fastidious bacteria.
Both are ideal for sampling aerobes; BD CultureSwab Plus™ is also appropriate for strict anaerobes. Systems consist of tube, transport medium, and swab with integral cap. Tubes are made of unbreakable polypropylene and include a preattached data label and a tamper-evident seal. Swabs are rayon and are designed with an extra long shaft. Various media are available for vaginal, skin, wound, nasopharyngeal, ear, nose, throat, and eye specimens, as well as male urethral sampling. Systems are also available with Cary-Blair agar for fecal specimens and with a sterile swab in an empty tube for general specimen collection and transport. Each swab package is flushed with nitrogen gas to assure freshness of the medium throughout the product shelf life.