BD™ Universal Viral Transport System, BD Medical
Supplier: BD Medical
BD™
220531 220528 220220 220220 220529
CA90006-624PK 211.63 CAD
10769-898 10769-896 CA90006-624 90006-624 10769-580
BD™ Universal Viral Transport System, BD Medical
Swabs Transport Swabs
The BD™ Universal Viral Transport System (UVT) is designed to transport viruses, chlamydiae, mycoplasmas, and ureaplasmas at room temperature. It is one of the only systems that may be stored and transported at 2 to 25 °C, all in one formulation.
  • No refrigeration requirements: System does not require refrigerated storage or transport on ice, decreasing the risk of specimen rejection and eliminating messy specimen transport
  • Reduced contamination: System features a unique blend of antimicrobics that target and suppress bacterial and fungal contamination without harming viruses, chlamydiae, mycoplasmas, or ureaplasmas
  • Long-term storage: System helps store samples for long periods by preserving viruses and chlamydiae, when stored frozen with a cryoprotectant