Multi-Disc™ Adapters for JS-4.0, JS-4.2, and JS-4.2A Rotors, Beckman Coulter®
Supplier: Beckman Coulter
339100 339101 339102 339103 339108 339109 341794 345386
CABK339100EA 391.06 CAD
CABK339100 CABK339101 CABK339102 CABK339103 CABK339108 CABK339109 CABK341794 CABK345386
Multi-Disc™ Adapters for JS-4.0, JS-4.2, and JS-4.2A Rotors, Beckman Coulter®
Centrifuges Rotors
Suitable for use with the JS-4.0 (BK339086) and JS-4.2/JS-4.2A (BK339080 series) swinging bucket rotors.
  • For swinging bucket rotors
  • Color-coded
  • Stackable
Color-coded adapters consist of multiple discs that may be stacked to the desired tube or bottle length. Maximum length for multiple-place adapters is 130mm; maximum length for single-place adapters is 160mm. Adapters are autoclavable.