Tube Racks for Aerosolve® Canisters, Beckman Coulter®
Supplier: Beckman Coulter
356985 358991 358992 359160 359161 359162 359163 359482 359483 359485 359486 359487 359488 359489 358993 359164 359484
CABK356985PK 611.04 CAD
CABK356985 CABK358991 CABK358992 CABK359160 CABK359161 CABK359162 CABK359163 CABK359482 CABK359483 CABK359485 CABK359486 CABK359487 CABK359488 CABK359489 CABK358993 CABK359164 CABK359484
Tube Racks for Aerosolve® Canisters, Beckman Coulter®
Racks Tube Racks
Molded plastic racks support tubes in Aerosolve® canisters.
  • Open-Rack Design for Quick Inspection
  • Molded Plastic Racks
  • Variety of Colors
The open-rack design permits quick inspection for possible broken or leaking tubes.