Modular Disk Adapters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter®
Supplier: Beckman Coulter
354511 359148 359149 359150 359152 359154 359155 359157 359469 359470 359472 359475 359476 359478 359151 359153 359156 359471 359473 359474 359477 359157
CABK354511EA 347.3 CAD
CABK354511 CABK359148 CABK359149 CABK359150 CABK359152 CABK359154 CABK359155 CABK359157 CABK359469 CABK359470 CABK359472 CABK359475 CABK359476 CABK359478 CABK359151 CABK359153 CABK359156 CABK359471 CABK359473 CABK359474 CABK359477 BK359157
Modular Disk Adapters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter®
Centrifuges Rotors
Adapters are for use with the SX4750 series, GH3.8, and JS-4.3 (BK362734) rotors.
  • Durable Modular Units
  • Thick Rubber Base
  • Autoclavable
  • Alternative Application: Storage Rack
For spinning 1.5 ml microcentrifuge tubes, a 26-hole adapter plate fits over the 12 mm blue adapters (BK359148) and (BK359469).
Durable modular units stack onto a thick rubber base to accommodate a wide variety of laboratory tubes. All units are autoclavable and can also be used as storage racks.