SP Bel-Art Spring Style Tubing Clamps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18213-0000 H18214-0000
63022-358PK 116.59 CAD
63022-358 63022-360
SP Bel-Art Spring Style Tubing Clamps, Bel-Art Products, a part of SP
Clamps Tubing and Sanitary Clamps
Perfect for all flexible tubing, this positive-action, self-closing clamp opens by squeezing the handles together.
  • Plastic and stainless steel construction
  • Self-closing clamp design
  • Easy to use
  • Suitable for use with all flexible tubing
Made of chemical and stain resistant Acetal plastic and stainless steel.