Sanitary Counter Brushes, Perfex
Supplier: Perfex
3050W 3050R 3050B 3050G 3050Y
22940-024EA 55.54 CAD
22940-024 22940-182 22940-186 22940-314 22940-316
Sanitary Counter Brushes, Perfex
Brushes
Sanitary Counter Brushes quickly remove fine dust, dirt and other debris from workstations, counter tops, and shelves without scattering particles.
  • Ideal for a variety of clean-up chores
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Poly block and fiber will not absorb bacteria, liquids or odors like nylon and natural fibers
  • Unaffected by water, grease, petroleum products, detergent, sanitizer and solvents
Medium-texture polypropylene fiber is heat-fused into impact-resistant polymer brush block, eliminating premature fiber fallout and areas for contaminants to collect.