VWR® Advanced 3750 Reciprocating Shakers
Supplier: VWR International
97007-742 10027-166
97007-742EA 6416.51 CAD
97007-742 10027-166
VWR® Advanced 3750 Reciprocating Shakers
Shakers Reciprocating Shakers
Digital reciprocating shakers are designed for a wide range of applications including solubility studies, extraction procedures, and cell cultures that require accurate and reproducible results.
  • Exceptional speed control, accuracy, and durability
  • Displays for speed and time
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
The shaking system delivers exceptional speed control, accuracy, and durability while continuously monitoring shaking speed and setpoint, even under changing loads.
Safety features include a built-in load sensor that detects unbalanced conditions and automatically reduces rpm or stops the operation to protect samples. A ramping feature slowly increases speed to the setpoint to avoid splashing. In timed mode, an audible alarm will sound when the timer reaches zero. Shakers can be run in cold rooms, incubators, and CO₂ environments from −10 to +60°C (−14 to +140°F). An RS-232 serial port is standard on these shakers.
Shakers can accommodate a variety of stainless steel platform options. Adjustable platforms are equipped with four cushioned rollers to hold most glassware securely. Universal platforms are suitable for mounting flask clamps and test tube racks. The culture platforms with mat are designed for slow speed applications that utilize low profile vessels. The separatory funnel platform can hold up to three 500 ml to 2 L samples at once. Standard test tube racks are made from PVC coated steel. Pivoting test tube racks are are made from polypropylene in a stainless steel holder.
The variable speed microprocessor control provides consistent, uniform mixing action. The microprocessor will display the last setpoint and will restart the unit if power is interrupted. Permanently lubricated ball bearings and a maintenance-free, brushless DC motor ensure reliable service in continuous operations. Touchpad controls feature easy-to-read, independent LEDs for speed and time, allowing operators to view all settings at once. The timer will display elapsed time or, when programmed to user defined limits, will shut off the unit. Last settings will be displayed even after the power has been shut off.
Certifications: RoHS certified. TUV listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information:
Shaker is supplied with a non-skid rubber mat to fit the 28×33 cm (11×13") tray, designed to secure Petri dishes and cell culture flasks. Note: tray and other accessories shown are not included and must be ordered separately. Unit also includes a 2.3 m (92") detachable 3-wire cord and plug. 230 V, 50/60 Hz models are also available; contact your Avantor sales representative for more information.