Owl™ Large Gel System, Models A1, A2 and A3, Thermo Scientific
Supplier: Thermo Fisher Scientific
A3-1 A2-BP
27372-038EA 1606.58 CAD
27372-038 27372-040 27372-070 27372-122 27373-304
Owl™ Large Gel System, Models A1, A2 and A3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Horizontal systems for detailed RNA/DNA analysis deliver consistent, quality results
A1 Model Systems feature a small footprint, conserving bench space. A2 Model Systems are wider than the A1 models, and can run up to 42 samples per comb position. They accomplish gel runs in 1–4 hours. A1 and A2 models are available with or without buffer exchange ports. A3 Model System is an extra-long apparatus designed especially for screening a large number of samples. The UVT gel tray for this system has multiple comb slots for more flexibility in comb placement. Gel tray fits 25- and 50-toothMore Product Information