Space-Saver Flammable Liquids Storage Cabinets, Vertical, Eagle Manufacturing
Supplier: Eagle Manufacturing
1923X 2310X
56610-250EA 6583.34 CAD
56610-250 56610-252
Space-Saver Flammable Liquids Storage Cabinets, Vertical, Eagle Manufacturing
Furniture Storage Cabinets
These space-saver cabinets fit where traditional 90.8 L (24 gal.) cabinets cannot, making it easy to safely store flammable liquids near work areas.
  • The self-closing door model is equipped with a fusible link that automatically closes the door if the link melts due to fire
  • Both models include three adjustable shelves
Cabinets are constructed of 18-gauge steel and painted with a caution yellow finish. The sides, top, and bottom are double-walled with 3.8 cm (³/₂") air space between walls. Additional features include grounding attachments, three point key-lock on doors, 5.1 cm (2") raised, leakproof doorsills, and four leveling feet.
Certifications: Flammable cabinets conform to OSHA flammable liquid storage requirements and NFPA Code 30 specifications.