Environmental Wastewater Certified Reference Materials
Supplier: SPEX CERTIPREP LLC
QCI-136 QCI-136C
NSQCI-136EA 225.96 CAD
NSQCI-136 NSQCI-136C
Environmental Wastewater Certified Reference Materials
Standards Environmental Analysis Standards
Validate analytical methods and evaluate the quality of your results with certified reference materials.
  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.
Certifications: Manufactured and certified under ISO 17034 accreditation. Meets your ISO 17025 requirements.