Corning® Aseptic Transfer Caps, Corning
Supplier: Corning
431445 431444 431447 431446 431449 431448
89089-896CS 1071.56 CAD
89089-896 89089-898 89089-900 89089-902 89089-904 89089-906
Corning® Aseptic Transfer Caps, Corning
Closures Safety Caps and Safety Waste Caps
Disposable transfer caps are for use with plastic Erlenmeyer flasks (see 29152-146 series)
Internal end of liquid transfer port rigid polypropylene dip tube angled to reach bottom of flask; external side has OD C-Flex tubing closed with female luer sealing cap.
Vent filter is 50 mm (2") in diameter, with a 0.2 µm pore PVDF membrane and barbed fitting. Internal end of liquid transfer port rigid polypropylene dip tube angled to reach bottom of flask; external side has OD C-Flex tubing closed with female luer sealing cap.
Delivery information: Transfer caps are supplied with a Sterility Assurance Level (SAL) of 10-6, and are double bagged.