Corning® Erlenmeyer Flask Caps, Polypropylene, Sterile, Corning
Supplier: Corning
431363 431339 431340 431364
89089-660CS 273.35 CAD
89089-660 89089-808 89089-820 89089-658
Corning® Erlenmeyer Flask Caps, Polypropylene, Sterile, Corning
Closures Bottle Closures
Made of PP.
Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.