Synthware West Condensers, Kemtech America
Supplier: Kemtech America
C151110 C150190 C159190 C159200 C154100 C151190 C154200 C154300
60000-968EA 150.48 CAD
60000-968 60000-970 60000-972 60000-974 60000-976 60000-978 60000-980 60000-982 60000-984
Synthware West Condensers, Kemtech America
Condensers
With narrow annular space for high cooling efficiency.
  • West style
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Serrated hose connection side arm
Narrow design increases flow rate of water, making for efficient cooling.