Axygen® 96-Well PCR Plates for MegaBACE™ Systems, Corning
Supplier: Corning
Axygen®
PCR-96-MB-C
47734-766CS 1111.19 CAD
47734-766
Axygen® 96-Well PCR Plates for MegaBACE™ Systems, Corning
Microplates PCR Plates
These plates are made of 99.9% pure virgin polypropylene and specifically designed for PCR applications
  • Max. RCF 4000 g
Optional AxyMat™ silicone sealing mat (10011-130) is autoclavable and freezable. Mats are pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. Alphanumeric grid system corresponds to 96-well plates.
They feature elevated chimney wells that are slightly taller and narrower than typical chimney wells, making these plates ideal for use with Amersham Biosciences MegaBACE™ DNA analysis systems. Plates are ideal for thermal transfer applications because their ultra-thin walls accelerate heat transfer to provide shorter cycles. Certified by lot number to be RNase-, DNase-, and pyrogen-free.
Ordering information: For sealing films, including film appropriate for Real Time PCR, see 10011-121 series.