Hand-Held Refractometers, ATAGO®
Supplier: Atago
2340-HSR-500 2350-R-5000
47752-824EA 2836.8 CAD
47752-824 47752-826
Hand-Held Refractometers, ATAGO®
Refractometers Handheld Refractometers
Model HSR-500 measures the full range of the Brix scale and has a +1.6 to –1.0 temperature compensation scale
Model R5000 is for easy measurement of a wide range of the refractive index and has a 5 to 40°C (41 to 104°F) thermometer. Both feature three-stage switching. Dimensions: 20Lx4Wx4Hcm (50.8x10.2x10.2"). Weight: 600g (21.2oz.).
Ordering information: Supplied with a one-year manufacturer’s warranty.