Axygen® 96-Well PCR Plates for ABI PRISM® 3100 Systems, Corning
Supplier: Corning
PCR-96M2-HS-C PCR-96M2-HS-R PCR-96M2-HS-Y
47743-953CS 1227.2 CAD
47743-953 47745-424 47745-430
Axygen® 96-Well PCR Plates for ABI PRISM® 3100 Systems, Corning
Microplates PCR Plates
These plates are made of 99.9% pure virgin polypropylene and are specifically designed for PCR and thermal transfer applications
  • Max. RCF 4000 g
Optional AxyMat™ silicone sealing mats are autoclavable and freezable. The standard AxyMat™ is pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. It also features an alphanumeric grid system that corresponds to 96-well plates. The AxyMat™ septa mat features individual septa to allow easy penetration and to prevent sample contamination and evaporation. It is designed specifically for use with the ABI PRISM® 3100 system.
Half-skirt design with A12 corner notch fits the Applied Biosystems ABI PRISM® 3100 Genetic Analyzer. Ultra-thin walls accelerate heat transfer to provide shorter cycles. All plates are certified by lot number to be RNase-, DNase-, and pyrogen-free.
Ordering information: Sealing mats sold separately. For sealing films, including film appropriate for Real Time PCR, see 10011-121 series.