Pesticide Surrogate Mix, EPA 8080, 8081, DCB and TCMX (2 Components), Restek
Supplier: RESTEK CORP
32000 32457
RK32000EA 90.59 CAD
RK32000 75835-018
Pesticide Surrogate Mix, EPA 8080, 8081, DCB and TCMX (2 Components), Restek
Standards Organic Standards
Contains two components, those are decachlorobiphenyl and 2,4,5,6-tetrachloro-m-xylene.
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.