CLP–Semi-Volatiles 3,3'-Dichlorobenzidine, Restek
Supplier: RESTEK CORP
31026 31835
RK31026EA 59.93 CAD
RK31026 75834-808
CLP–Semi-Volatiles 3,3'-Dichlorobenzidine, Restek
Standards Environmental Analysis Standards
Standards are packaged at a concentration of 2000 µg/mL in a 1 mL ampule.
  • Mixture contains 3,3'-Dichlorobenzidine
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).