Environmental standards, Semi-Volatiles (SV) Internal Standard Mix, Restek
Supplier: RESTEK CORP
31006 31206 31006-15 31206-15 31206-25 31006-25
RK31006EA 247.5 CAD
RK31006 RK31206 75834-596 75834-632 75834-634 75835-616
Environmental standards, Semi-Volatiles (SV) Internal Standard Mix, Restek
Standards Environmental Analysis Standards
Standards are packaged in methylene chloride in a 1 mL ampoule.
  • High purity for consistent results
  • Free quality assurance data package available online
  • Highest concentrations commercially available.
Mixture contains Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, and Phenanthrene
US Environmental Protection Agency (EPA) Method 8270D outlines the analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices, using gas chromatography/mass spectrometry (GC/MS). Update IVA of the third edition of SW-846—Test Methods for Evaluating Solid Waste, Physical/Chemical Methods—includes EPA Method 8270D, in which there were no major revisions from EPA Method 8270C.
Delivery information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).