B/N Surrogate Mix (4/89 SOW), Restek
Supplier: RESTEK CORP
31024 31062 31086 31024-25 31062-15 33028 31024-15
RK31024EA 68.73 CAD
RK31024 RK31062 RK31086 75834-598 75834-608 75835-104 75835-620
B/N Surrogate Mix (4/89 SOW), Restek
Standards Environmental Analysis Standards
Base/neutrals surrogate standards (3 components) are packaged in methylene chloride in an ampoule.
  • Convenient 1 mL 5 mL and 10 mL package sizes
  • Highest concentrations commercially available
  • High purity for consistent results
Mixture contains 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14
Delivery information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).