Acid Surrogate Mix (4/89 SOW), Restek
Supplier: RESTEK CORP
31063 31087 31025 31025-15 31025-25 31063-15 33029
RK31063EA 93.22 CAD
RK31063 RK31087 RK31025 75834-600 75834-602 75834-610 75835-108
Acid Surrogate Mix (4/89 SOW), Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol in an ampule.
  • Convenient 1 mL, 5 mL and 10 mL package sizes
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
  • Mixture contains 2-Fluorophenol, Phenol-d6, and 2,4,6-Tribromophenol
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).