Effluent Level Inorganics Wastewater Standard, NSI
Supplier: SPEX CERTIPREP LLC
QCI-072 QCI-072C
NSQCI-072EA 184.65 CAD
NSQCI-072 NSQCI-072C
Effluent Level Inorganics Wastewater Standard, NSI
Standards Environmental Analysis Standards
Whole volume standard
Standard contains 25 mg/L of COD, 1 mg/L of NH3-N, 2 mg/L of N-NO3, 1 mg/L of p-PO4, 25 mg/L of SO4, and 8 mg/L of TOC.
Includes CoA listing true value and acceptance limits. Supplied in HDPE bottle.
Certifications: NIST traceable