Paramount® and Paramount 360® Ductless Enclosures, Labconco®
Supplier: Labconco
6963200 6963220 6963300 6963301 6963302 6963320 6963321 6963322 6963400 6963401 6963402 6963403 6963420 6963421 6963422 6963423 6963501 6963503 6963521 6963523 6963601 6963603 6963621 6963623 6938200 6938201 6938202 6938203 6938100 6938100 6938200 6938201 6938202 6938203 6963302
97007-602EA 11834.2 CAD
97007-602 97007-604 97007-606 97007-608 97007-610 97007-612 97007-614 97007-616 97007-618 97007-620 97007-622 97007-624 97007-626 97007-628 97007-630 97007-632 97007-634 97007-636 97007-638 97007-640 97007-642 97007-644 97007-646 97007-648 97007-650 97007-652 97007-654 97007-656 97007-662 470203-232 470203-234 470203-236 470203-238 470203-240 470203-242
Paramount® and Paramount 360® Ductless Enclosures, Labconco®
Fume Hoods and Enclosures Ductless Fume Hoods
Self-contained ductless enclosures allow safe handling of organic, formaldehyde, acid gas, sulfur compounds, or ammonia chemical fumes and vapors.
  • Upper Containment Sash Foil
  • Upper Dilution Air Supply
  • Slotted Rear Baffle
  • Enclosures Feature 360° Visibility
  • Feet Can Be Replaced with Casters for Mobility
Enclosures feature an upper containment sash foil, upper dilution air supply, and a slotted rear baffle that promotes horizontal laminar airflow by drawing vapors and fumes to the back of the enclosure without causing turbulence that can impede containment
Stands have a depth of 87 cm (34¼"). The built-in hydraulic lift is operated by remote control and provides work surface heights from 43.8–93.3 cm (17¼–36¾") and overall enclosure height as low as 193.5 cm (76¼"). The supported enclosure can be positioned for persons of different statures, wheelchair-bound users, transport through doorways, and easier access to upper components. Stands include 12.7 cm (5") diameter, toe-locking polyurethane casters with bearings for quiet operation. Black, chemical-resistant epoxy work surfaces are dished to contain spills and contoured to fit enclosures. Stainless steel work surfaces feature a 2.5 cm (1") deep-front spill trough. Air inside the enclosures passes through carbon filters, and in some cases additional HEPA particulate filters, before being exhausted back into the laboratory. No outside ducting is required.
Tempered safety glass front sash and sides, epoxy-coated aluminum frame, steel rear baffle, and plenum provide excellent visibility, ambient light, and chemical resistance. Paramount 360 enclosures feature 360° visibility. Ergonomic angled sash allows a closer view of work area, reduces glare, and provides a more comfortable position for sitting or standing operation. Advanced motor technology uses less energy and runs quieter than comparable models. Containment-enhancing components ensure that no vapors enter the breathing zone for a safe work environment. An organic vapor sensor detects chemicals breaking through the filters, ensuring operator protection. Digital display shows airflow and filter status, and touchpad controls enclosure operation.
Enclosures meet SEFA 1-2005, ASHRAE 110-95, and ANSI z9.5-2003 standards. CAN/CSA C22.2 No. 1010.1 compliant. UL 3101-1/61010-1 compliant. 115V models are also ETL and ETL-C listed. 230V models are also CE marked.
Certifications: Enclosures meet SEFA 1-2005, ASHRAE 110-95, and ANSI z9.5-2003 standards. CAN/CSA C22.2 No. 1010.1 compliant. UL 3101-1/61010-1 compliant. 115V models are also ETL and ETL-C listed. 230V models are also CE marked.
Ordering information: All models require a base stand, work surface, and filters in order to operate (all sold separately). Filters must be selected according to individual application requirements and can be stacked two deep. Only two types of filters can be used together or both can be of the same type.