Basic™ 47 and Basic 70 Laboratory Hoods, Labconco®
Supplier: Labconco
2246601 2246603 2246604 2246602 2247401 2247301 2246600 2246605 2246606 2246300 2246301 2246400 2246401 2246500 2246501 2246706 2246705 2246704 2246703 2246702 2246701 2246700 2247300 2247400 2247500 2247501
30140-200EA 9898.7 CAD
30140-200 30140-202 30140-204 30140-206 30140-208 30140-210 30140-214 30140-216 30140-218 30140-220 30140-222 30140-224 30140-226 30140-228 30140-230 30140-232 30140-234 30140-236 30140-238 30140-240 30140-242 30140-244 30152-518 30152-562 30152-610 30152-612
Basic™ 47 and Basic 70 Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
The Basic™ hood is ideal for rooms with ceilings below 2.4 m, 8" and for shallow countertops.
  • Blowers Available
  • Safety Glass
  • Steel Construction
  • Vapor-Proof, Incandescent Light Fixture
The hood is furnished with a 0.5 cm (3/16") thick safety glass and a 25.4 cm (10") dia
The hood is available with or without a blower, which can be mounted in or above the hood or on the roof. A 1/3 hp motor is included with Basic 47 hoods that have a blower (except 30152-610 and 612, which have a 1/2 hp motor). Basic 70 hoods with a blower have a 3/4 hp motor.