Purifier® Class I Safety Enclosures, Labconco®
Supplier: Labconco
3980201 3980221 3980301 3980321 3980203 3980223 3980303 3980323 3980401 3980403 3980421 3980423
82018-092EA 11747.3 CAD
82018-092 82018-094 82018-096 82018-098 82018-156 82018-158 82018-160 82018-162 82018-164 82018-166 82018-168 82018-170
Purifier® Class I Safety Enclosures, Labconco®
Fume Hoods and Enclosures Laminar Flow Cabinets
These enclosures provide practical, economical protection of operator and environment for applications involving biohazardous material and toxic particulates
During operation, room air is drawn from around the operator, through the work area, and into the HEPA filter. A 99.99% efficient HEPA filter removes all particles >=0.3µm. The patented Clean-Sweep™ airfoil allows air to sweep the work surface, while the patented rear baffle with zones of perforations promotes horizontal laminar airflow to maximize containment. An upper containment sash foil bleeds air into the enclosure to directMore Product Information