Purifier® HEPA-Filtered Enclosures, Labconco®
Supplier: Labconco
3980200 3980220 3980300 3980320 3980202 3980222 3980302 3980322 3980400 3980402 3980420 3980422
82018-100EA 10878.3 CAD
82018-100 82018-102 82018-104 82018-106 82018-172 82018-174 82018-176 82018-178 82018-180 82018-182 82018-184 82018-186
Purifier® HEPA-Filtered Enclosures, Labconco®
Fume Hoods and Enclosures Laminar Flow Cabinets
These enclosures provide practical, economical protection of operator and environment for applications that generate fine dusts or aerosols but do not provide product protection
During operation, room air is drawn from around the operator, through the work area, and into the HEPA filter. A 99.99% efficient HEPA filter removes all particles >=0.3µm. The patented Clean-Sweep™ airfoil allows air to sweep the work surface, while the patented rear baffle with zones of perforations promotes horizontal laminar airflow to maximize containment. An upper containment sash foil bleeds air into theMore Product Information