XPert™ Balance Enclosures, Labconco®
Supplier: Labconco
3930201 3930202 3930221 3930222 3930301 3930302 3930321 3930322 3930401 3930402 3930421 3930422 3930231 3930331 3930431 3930232 3930332 3930432 3930241 3930242 3930341 3930342 3930441 3930442 3930200 3930230 3930300 3930330 3930400 3930430 3971500 3971501 3971502 3971521 3971522 3971530 3971531 3971532 3971541 3971542 3971560 3971561 3971562 3971571 3971572 3971600 3971601 3971602 3971621 3971622 3971630 3971631 3971632 3971641 3971642 3971660 3971661 3971662 3971671 3971672 3971800 3971801 3971802 3971821 3971822 3971830 3971831 3971832 3971841 3971842 3971860 3971861 3971862 3971871 3971872
82010-644EA 7915.8 CAD
82010-644 82010-646 82010-648 82010-650 82010-652 82010-654 82010-656 82010-658 82010-660 82010-662 82010-664 82010-666 89003-066 89003-068 89003-070 89004-440 89004-442 89004-444 89004-446 89004-448 89004-450 89004-452 89004-454 89004-456 89004-458 89004-460 89004-462 89004-464 89004-466 89004-468 93000-414 93000-416 93000-418 93000-420 93000-422 93000-424 93000-426 93000-428 93000-430 93000-432 93000-434 93000-436 93000-438 93000-440 93000-442 93000-444 93000-446 93000-448 93000-450 93000-452 93000-454 93000-456 93000-458 93000-460 93000-462 93000-464 93000-466 93000-468 93000-470 93000-472 93000-474 93000-476 93000-478 93000-480 93000-482 93000-484 93000-486 93000-488 93000-490 93000-492 93000-494 93000-496 93000-498 93000-500 93000-502
XPert™ Balance Enclosures, Labconco®
Fume Hoods and Enclosures Balances Enclosures
These low-profile enclosures provide user protection by keeping powders, particulates, and fumes contained during weighing procedures
Sturdy glacier white and gray, dry powder epoxy-coated aluminum frame and steel rear plenum and baffle dissipate static charge. Tempered safety glass front sash, sides, and top offer excellent visibility, ambient light, and protection. All models with interior heights of 56.5cm (221/4") have an operating sash height of 20.3cm (8"). The 0.6m (2'), 0.9m (3'), and 1.2m (4') models with interior heights of 80cm (311/2") have an operating sash height of 25.4cm (1More Product Information