VWR® Bath Fluids
Supplier: VWR International
13270-792 13270-794 13272-034
13270-792EA 3370.34 CAD
13270-792 13270-794 13272-034
VWR® Bath Fluids
Cleaning Agents Water Bath Protection Reagents
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
  • Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
Packaging: Bottles are packaged in a carton for shipment.