Corning® Thermowell® GOLD 384-Well PCR Microplates, Corning
Supplier: Corning
Corning®
3757
33502-106CS 695.18 CAD
33502-106
Corning® Thermowell® GOLD 384-Well PCR Microplates, Corning
Microplates PCR Plates
PP, non sterile. Compatible with a full range of PCR, sequencing and Real-Time PCR equipment.
Thermowell® GOLD 384-well PCR plates feature exceptional dimensional stability following thermocycling, and are fully compatible with automation, commonly used thermal cyclers, and Applied Biosystems® sequencing adapters.
Ordering information: Sealing tape and sealing mats sold separately.