Corning® 96-Well Polypropylene Tubes, Corning
Supplier: Corning
4401 4408 4410 4411 4412 4413 4418
29442-602CS 133.97 CAD
29442-602 29442-604 29442-606 29442-608 29442-610 29442-612 29442-614
Corning® 96-Well Polypropylene Tubes, Corning
Tubes Culture Tubes
PP tubes, PE tube caps.
  • Composed of 96 PP tubes in a standard microplate format
  • 1.2 ml tubes available individually or in strips of 8 tubes
  • PE tube caps available in 8-cap strips
Packaging: Supplied bagged or racked.