Stripwell™ 96-well Standard Polystyrene Microplates, Corning®
Supplier: Corning
9102 2592 2593 3923 3924 2580
29442-072CS 926.63 CAD
29442-072 29442-302 29442-304 29444-024 29444-026 29444-028 29444-030 10068-004
Stripwell™ 96-well Standard Polystyrene Microplates, Corning®
Microplates Immunology Microplates and Strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation